missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat IGSF8/CD316 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB31171-SP
This item is not returnable.
View return policy
Description
IGSF8/CD316 Monoclonal antibody specifically detects IGSF8/CD316 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Specifications
| IGSF8/CD316 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| CD316, CD316 antigen, CD81P3, CD81P3LIR-D1, EWI2, EWI-2, EWI2CD81 partner 3, Glu-Trp-Ile EWI motif-containing protein 2, IgSF8, immunoglobulin superfamily, member 8, KCT4, KCT-4, Keratinocytes-associated transmembrane protein 4, PGRL, PGRLimmunoglobulin superfamily member 8 | |
| Human embryonic kidney cell, HEK293-derived human IGSF8/CD316, Arg28-Thr579, Accession # Q969P0-1 | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG |
| Western Blot, Flow Cytometry, CyTOF | |
| 2587G | |
| Western Blot 2 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/106 cells, CyTOF-ready | |
| Q969P0-1 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 93185 | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction