missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat GIT1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£167.00 - £361.00
Specifications
| Antigen | GIT1 |
|---|---|
| Clone | 924640 |
| Dilution | Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready |
| Applications | Western Blot, Immunohistochemistry, Flow Cytometry, CyTOF |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18732569
![]() |
R&D Systems
MAB8508-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18752708
![]() |
R&D Systems
MAB8508 |
100 μg |
£361.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GIT1 Monoclonal specifically detects GIT1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready.Specifications
| GIT1 | |
| Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ARF GAP GIT1, ARF GTPase-activating protein GIT1, CAT1, CAT-1, Cool-associated and tyrosine-phosphorylated protein 1, G protein-coupled receptor kinase interacting ArfGAP 1, G protein-coupled receptor kinase interactor 1, G protein-coupled receptor kinase-interactor 1, GRK-interacting protein 1 | |
| GIT1 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 924640 | |
| Western Blot, Immunohistochemistry, Flow Cytometry, CyTOF | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| Q9Y2X7 | |
| 28964 | |
| E. coli-derived recombinant human GIT1 Ser485-Asp636 Accession # Q9Y2X7 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human GIT1 in direct ELISAs and human, mouse, and rat GIT1 in Western blots. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title