missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat GIT1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB8508-SP
This item is not returnable.
View return policy
Description
GIT1 Monoclonal specifically detects GIT1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready.
Specifications
| GIT1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ARF GAP GIT1, ARF GTPase-activating protein GIT1, CAT1, CAT-1, Cool-associated and tyrosine-phosphorylated protein 1, G protein-coupled receptor kinase interacting ArfGAP 1, G protein-coupled receptor kinase interactor 1, G protein-coupled receptor kinase-interactor 1, GRK-interacting protein 1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 28964 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot, Immunohistochemistry, Flow Cytometry, CyTOF | |
| 924640 | |
| Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| Q9Y2X7 | |
| GIT1 | |
| E. coli-derived recombinant human GIT1 Ser485-Asp636 Accession # Q9Y2X7 | |
| 25 μg | |
| Primary | |
| Detects human GIT1 in direct ELISAs and human, mouse, and rat GIT1 in Western blots. | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction