missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat EG-VEGF/PK1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£146.00 - £452.00
Specifications
| Antigen | EG-VEGF/PK1 |
|---|---|
| Clone | 239727 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18793427
![]() |
R&D Systems
MAB2100-SP |
25 μg |
£146.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18162561
|
R&D Systems
MAB2100 |
500 μg |
£452.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
EG-VEGF/PK1 Monoclonal specifically detects EG-VEGF/PK1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
| EG-VEGF/PK1 | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| Q8R414 | |
| 84432 | |
| Mouse myeloma cell line NS0-derived recombinant rat EG-VEGF Ala20-Phe105 (predicted) Accession # Q8R414 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human, mouse, and rat EG-VEGF in direct ELISAs and Western blots. In direct ELISAs, 100% cross-reactivity with recombinant human EG-VEGF and recombinant mouse EG-VEGF is observed. |
| 239727 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EGVEGF, EG-VEGF, EGVEGFblack mamba toxin-related protein, Endocrine-gland-derived vascular endothelial growth factor, Mambakine, PK1mambakine, PRK1prokineticin-1, PROK1, prokineticin 1 | |
| PROK1 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title