missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat EG-VEGF/PK1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB2100-SP
This item is not returnable.
View return policy
Description
EG-VEGF/PK1 Monoclonal specifically detects EG-VEGF/PK1 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| EG-VEGF/PK1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q8R414 | |
| PROK1 | |
| Mouse myeloma cell line NS0-derived recombinant rat EG-VEGF Ala20-Phe105 (predicted) Accession # Q8R414 | |
| 25 μg | |
| Primary | |
| Detects human, mouse, and rat EG-VEGF in direct ELISAs and Western blots. In direct ELISAs, 100% cross-reactivity with recombinant human EG-VEGF and recombinant mouse EG-VEGF is observed. | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| 239727 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EGVEGF, EG-VEGF, EGVEGFblack mamba toxin-related protein, Endocrine-gland-derived vascular endothelial growth factor, Mambakine, PK1mambakine, PRK1prokineticin-1, PROK1, prokineticin 1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 84432 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction