missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat CaM Kinase II Pan Specific Antibody, R&D Systems™

Mouse Monoclonal Antibody
£167.00 - £362.00
Specifications
| Antigen | CaMKinaseII |
|---|---|
| Clone | 990714 |
| Dilution | Western Blot 1μg/mL, Immunohistochemistry 0.5-25μg/mL, Immunocytochemistry 5-25μg/mL |
| Applications | Immunocytochemistry, Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18093332
![]() |
R&D Systems
MAB7280-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18051102
|
R&D Systems
MAB7280-100 |
100 μg |
£362.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CaM Kinase II Monoclonal specifically detects CaM Kinase II in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.Specifications
| CaMKinaseII | |
| Western Blot 1μg/mL, Immunohistochemistry 0.5-25μg/mL, Immunocytochemistry 5-25μg/mL | |
| Monoclonal | |
| Supernatant | |
| Human, Mouse, Rat | |
| Q13555 | |
| E. coli-derived recombinant human CaM Kinase II gamma Ala448-Gln558 Accession # Q13555 | |
| Primary | |
| Reconstitute at 0.5mg/mL in sterile PBS. | |
| Detects human CaM Kinase II gamma in direct ELISAs. Detects human CaM Kinase II alpha , beta , gamma , and delta in Western blots. Detects human, mouse, and rat CaM Kinase II in Western blots. |
| 990714 | |
| Immunocytochemistry, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
| 816 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title