Learn More
Human/Mouse/Rat CaM Kinase II Pan Specific Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB7280-100
This item is not returnable.
View return policy
Description
CaM Kinase II Monoclonal specifically detects CaM Kinase II in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.
Specifications
| CaMKinaseII | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| Primary | |
| Detects human CaM Kinase II gamma in direct ELISAs. Detects human CaM Kinase II alpha , beta , gamma , and delta in Western blots. Detects human, mouse, and rat CaM Kinase II in Western blots. | |
| Human, Mouse, Rat | |
| Supernatant |
| Immunocytochemistry, Immunohistochemistry, Western Blot | |
| 990714 | |
| Western Blot 1μg/mL, Immunohistochemistry 0.5-25μg/mL, Immunocytochemistry 5-25μg/mL | |
| Q13555 | |
| E. coli-derived recombinant human CaM Kinase II gamma Ala448-Gln558 Accession # Q13555 | |
| 100 μg | |
| 816 | |
| Reconstitute at 0.5mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |