missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Activin A beta A subunit Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 9 publications
£187.00 - £1795.00
Specifications
| Antigen | Activin A |
|---|---|
| Clone | 69403 |
| Dilution | Neutralization 0.02-0.06 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Applications | Neutralization, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18718548
![]() |
R&D Systems
MAB3381-SP |
25 μg |
£187.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18727739
![]() |
R&D Systems
MAB3381-100 |
100 μg |
£447.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18788039
![]() |
R&D Systems
MAB3381-500 |
500 μg |
£1795.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Activin A Monoclonal specifically detects Activin A in Human, Mouse, Rat samples. It is validated for Neutralization, ELISA Capture (Matched Antibody Pair).Specifications
| Activin A | |
| Neutralization 0.02-0.06 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human, Mouse, Rat | |
| INHBA | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 69403 | |
| Neutralization, ELISA | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Chinese hamster ovary cell line CHO-derived recombinant human Activin A | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects recombinant human Activin A in ELISAs. Also recognizes Activin A precursor. The amino acid sequences of mature beta A subunits from human, mouse and rat are identical. Therefore, this antibody will also detect the mature beta A subunit from mouse and rat. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title