missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Activin A beta A subunit Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 9 publications
Brand: R&D Systems MAB3381-500
This item is not returnable.
View return policy
Description
Activin A Monoclonal specifically detects Activin A in Human, Mouse, Rat samples. It is validated for Neutralization, ELISA Capture (Matched Antibody Pair).
Specifications
| Activin A | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| INHBA | |
| Chinese hamster ovary cell line CHO-derived recombinant human Activin A | |
| 500 μg | |
| Primary | |
| Detects recombinant human Activin A in ELISAs. Also recognizes Activin A precursor. The amino acid sequences of mature beta A subunits from human, mouse and rat are identical. Therefore, this antibody will also detect the mature beta A subunit from mouse and rat. | |
| Human, Mouse, Rat | |
| Supernatant |
| Neutralization, ELISA | |
| 69403 | |
| Neutralization 0.02-0.06 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P18075 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 655 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction