missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse PKC beta 2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£157.00 - £337.00
Specifications
| Antigen | PKC beta 2 |
|---|---|
| Clone | 469827 |
| Dilution | Western Blot 1 ug/mL, Immunohistochemistry 5-25 ug/mL |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18736558
![]() |
R&D Systems
MAB43781-SP |
25 μg |
£157.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18744748
![]() |
R&D Systems
MAB43781-100 |
100 μg |
£337.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PKC beta 2 Monoclonal specifically detects PKC beta 2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| PKC beta 2 | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| 5579 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 469827 | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human, Mouse | |
| P05771 | |
| E. coli-derived recombinant human PKC beta 2 Lys607-Ser673 Accession # P05771 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human PKC beta 2 in direct ELISAs. Detects human and mouse PKC beta 2 in Western blots. |
RUO
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title