missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse PKC beta 2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB43781-SP
This item is not returnable.
View return policy
Description
PKC beta 2 Monoclonal specifically detects PKC beta 2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| PKC beta 2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5579 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, Immunohistochemistry | |
| 469827 | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-25 ug/mL | |
| P05771 | |
| E. coli-derived recombinant human PKC beta 2 Lys607-Ser673 Accession # P05771 | |
| 25 μg | |
| Primary | |
| Detects human PKC beta 2 in direct ELISAs. Detects human and mouse PKC beta 2 in Western blots. | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
RUO
Spot an opportunity for improvement?Share a Content Correction