missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Orexin A/Hypocretin-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£184.00 - £1785.00
Specifications
| Antigen | Orexin A/Hypocretin-1 |
|---|---|
| Clone | 97505 |
| Dilution | Immunohistochemistry 5-25 ug/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18759118
![]() |
R&D Systems
MAB763-SP |
25 μg |
£184.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18799427
![]() |
R&D Systems
MAB763-100 |
100 μg |
£439.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18737639
![]() |
R&D Systems
MAB763-500 |
500 μg |
£1785.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Orexin A/Hypocretin-1 Monoclonal antibody specifically detects Orexin A/Hypocretin-1 in Human, Mouse samples. It is validated for Immunohistochemistry.Specifications
| Orexin A/Hypocretin-1 | |
| Immunohistochemistry 5-25 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human, Mouse, Rat | |
| O43612 | |
| 3060 | |
| KLH-coupled human Orexin A/Hypocretin-1 synthetic peptide Accession # O43612 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human and mouse Orexin A/Hypocretin-1. In dot blots, no cross-reactivity with recombinant human Orexin B is observed. |
| 97505 | |
| Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| HCRT, Hcrt1, hypocretin (orexin) neuropeptide precursor, Hypocretin1, Hypocretin-1, NRCLP1, orexin, OXHcrt, PPORX, PPOXHypocretin | |
| HCRT | |
| IgG1 | |
| Protein A or G purified from ascites | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title