missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Orexin A/Hypocretin-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB763-500
This item is not returnable.
View return policy
Description
Orexin A/Hypocretin-1 Monoclonal antibody specifically detects Orexin A/Hypocretin-1 in Human, Mouse samples. It is validated for Immunohistochemistry.
Specifications
| Orexin A/Hypocretin-1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| HCRT, Hcrt1, hypocretin (orexin) neuropeptide precursor, Hypocretin1, Hypocretin-1, NRCLP1, orexin, OXHcrt, PPORX, PPOXHypocretin | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 3060 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Immunohistochemistry | |
| 97505 | |
| Immunohistochemistry 5-25 ug/mL | |
| O43612 | |
| HCRT | |
| KLH-coupled human Orexin A/Hypocretin-1 synthetic peptide Accession # O43612 | |
| 500 μg | |
| Primary | |
| Detects human and mouse Orexin A/Hypocretin-1. In dot blots, no cross-reactivity with recombinant human Orexin B is observed. | |
| Human, Mouse, Rat | |
| Ascites |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction