missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Olig3 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
£170.00 - £390.00
Specifications
| Antigen | Olig3 |
|---|---|
| Clone | 257934 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot, Immunohistochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18703419
![]() |
R&D Systems
MAB2456-SP |
25 μg |
£170.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18131801
|
R&D Systems
MAB2456 |
100 μg |
£390.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Olig3 Monoclonal specifically detects Olig3 in Human, Mouse samples. It is validated for Western Blot.Specifications
| Olig3 | |
| LYOPH | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human, Mouse | |
| Q7RTU3 | |
| 167826 | |
| E. coli-derived recombinant human Olig3 Met1-Lys272 Accession # Q7RTU3 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human and mouse Olig3 in Western blots. In Western blots, no cross-reactivity with recombinant human (rh) Olig1 or rhOlig2 is observed. |
| 257934 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Bhlhb7, BHLHE20, bHLHe20oligo3, Class B basic helix-loop-helix protein 7, Class E basic helix-loop-helix protein 20, Oligo3, oligodendrocyte transcription factor 3 | |
| OLIG3 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title