missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Olig3 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
Brand: R&D Systems MAB2456-SP
This item is not returnable.
View return policy
Description
Olig3 Monoclonal specifically detects Olig3 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| Olig3 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q7RTU3 | |
| OLIG3 | |
| E. coli-derived recombinant human Olig3 Met1-Lys272 Accession # Q7RTU3 | |
| 25 μg | |
| Primary | |
| Detects human and mouse Olig3 in Western blots. In Western blots, no cross-reactivity with recombinant human (rh) Olig1 or rhOlig2 is observed. | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 257934 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Bhlhb7, BHLHE20, bHLHe20oligo3, Class B basic helix-loop-helix protein 7, Class E basic helix-loop-helix protein 20, Oligo3, oligodendrocyte transcription factor 3 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 167826 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction