missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Oct-3/4 PE-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 9 publications
£254.00 - £453.00
Specifications
| Antigen | Oct-3/4 |
|---|---|
| Clone | 240408 |
| Dilution | Intracellular Staining by Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18779658
![]() |
R&D Systems
IC1759P-025 |
25 Tests |
£254.00
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18703947
![]() |
R&D Systems
IC1759P-100 |
100 Tests |
£453.00
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Oct-3/4 Monoclonal specifically detects Oct-3/4 in Human, Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
| Oct-3/4 | |
| Intracellular Staining by Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| 5460 | |
| E. coli-derived recombinant human Oct-3/4 Met1-Asn265 (Met262Leu) Accession # Q01860 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
| 240408 | |
| Flow Cytometry | |
| R-PE | |
| Rat | |
| Human, Mouse | |
| Q01860 | |
| POU5F1 | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects human Oct-3/4 in Western blots and detects mouse Oct-3/4 in flow cytometry. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title