missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Oct-3/4 PE-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 9 publications
Brand: R&D Systems IC1759P-100
This item is not returnable.
View return policy
Description
Oct-3/4 Monoclonal specifically detects Oct-3/4 in Human, Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| Oct-3/4 | |
| Monoclonal | |
| R-PE | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| POU5F1 | |
| E. coli-derived recombinant human Oct-3/4 Met1-Asn265 (Met262Leu) Accession # Q01860 | |
| 100 Tests | |
| Primary | |
| Detects human Oct-3/4 in Western blots and detects mouse Oct-3/4 in flow cytometry. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2b |
| Flow Cytometry | |
| 240408 | |
| Intracellular Staining by Flow Cytometry 10 uL/10^6 cells | |
| Q01860 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5460 | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction