missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse MOG Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications
£171.00 - £371.00
Specifications
| Antigen | MOG |
|---|---|
| Clone | 296829 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18751448
![]() |
R&D Systems
MAB2439-SP |
25 μg |
£171.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18119060
|
R&D Systems
MAB2439 |
100 μg |
£371.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MOG Monoclonal specifically detects MOG in Human, Mouse samples. It is validated for Western Blot.Specifications
| MOG | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Rat | |
| Human, Mouse | |
| Q61885 | |
| 4340 | |
| Mouse myeloma cell line NS0-derived recombinant mouse MOG Gly29-Gly153 Accession # Q61885 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human and mouse MOG in direct ELISAs and Western blots. |
| 296829 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| MGC26137, MOG Ig-AluB, MOGIG2, myelin oligodendrocyte glycoprotein, myelin-oligodendrocyte glycoprotein | |
| MOG | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title