missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse MOG Antibody, R&D Systems™
Rat Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB2439
This item is not returnable.
View return policy
Description
MOG Monoclonal specifically detects MOG in Human, Mouse samples. It is validated for Western Blot.
Specifications
| MOG | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q61885 | |
| MOG | |
| Mouse myeloma cell line NS0-derived recombinant mouse MOG Gly29-Gly153 Accession # Q61885 | |
| 100 μg | |
| Primary | |
| Detects human and mouse MOG in direct ELISAs and Western blots. | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| 296829 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| MGC26137, MOG Ig-AluB, MOGIG2, myelin oligodendrocyte glycoprotein, myelin-oligodendrocyte glycoprotein | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4340 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction