missing translation for 'onlineSavingsMsg'
Learn More

Human/Mouse FGF-21 Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody has been used in 2 publications

£167.00 - £420.00

Specifications

Antigen FGF-21
Clone 315901
Concentration LYOPH
Dilution Western Blot 1 ug/mL
Applications Western Blot
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18796959
GREENER_CHOICE
View Documents
R&D Systems
MAB25371-SP
25 μg
£167.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18171941
GREENER_CHOICE
View Documents
R&D Systems
MAB25371
100 μg
£420.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

FGF-21 Monoclonal specifically detects FGF-21 in Human, Mouse samples. It is validated for Western Blot.
Specifications

Specifications

FGF-21
LYOPH
Western Blot
Unconjugated
Mouse
Human, Mouse
Q9NSA1
26291
E. coli-derived recombinant human FGF-21 His29-Ser209 Accession # Q9NSA1
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects human and mouse FGF-21 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human (rh) FGF-12, -13, -16, -17, -19, -20, -23, rhFGF basic, or rhFGF acidic.
315901
Western Blot 1 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
FGF21, FGF-21, fibroblast growth factor 21
FGF21
IgG1
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.