missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse FGF-21 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB25371
This item is not returnable.
View return policy
Description
FGF-21 Monoclonal specifically detects FGF-21 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| FGF-21 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q9NSA1 | |
| FGF21 | |
| E. coli-derived recombinant human FGF-21 His29-Ser209 Accession # Q9NSA1 | |
| 100 μg | |
| Primary | |
| Detects human and mouse FGF-21 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human (rh) FGF-12, -13, -16, -17, -19, -20, -23, rhFGF basic, or rhFGF acidic. | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| 315901 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FGF21, FGF-21, fibroblast growth factor 21 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 26291 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction