missing translation for 'onlineSavingsMsg'
Learn More

Human/Mouse Dkk-1 Antibody, R&D Systems™
GREENER_CHOICE

Product Code. 18799067
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18799067 25 μg 25µg
18001762 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18799067 Supplier R&D Systems Supplier No. AF1096SP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 15 publications

Produced in goats immunized with purified, insect cell line Sf 21-derived, recombinant human Dickkopf homolog-1 (rhDkk-1). Human Dkk-1 specific IgG was purified by human Dkk-1 affinity chromatography.

Dkk-1 Polyclonal specifically detects Dkk-1 in Human, Mouse samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction, Knockout Validated.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Dkk-1
Applications Western Blot, Blockade of Receptor-ligand Interaction, Immunoassay
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Blockade of Receptor-ligand Interaction, Knockout Validated
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. with No Preservative
Gene Accession No. O94907
Gene Alias dickkopf (Xenopus laevis) homolog 1, dickkopf homolog 1 (Xenopus laevis), dickkopf related protein-1, Dickkopf-1, dickkopf-related protein 1, Dkk1, DKK-1, hDkk-1, SKdickkopf-1 like
Gene Symbols DKK1
Host Species Goat
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Dkk-1 Thr32-His266 Accession # O94907
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 22943
Test Specificity Detects human and mouse Dkk-1 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) Dkk-2, and rhDkk-4 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.