missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Chicken RGM-A Antibody, R&D Systems™

Rat Monoclonal Antibody
£157.00 - £346.00
Specifications
| Antigen | RGM-A |
|---|---|
| Clone | 275407 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18712539
![]() |
R&D Systems
MAB2458-SP |
25 μg |
£157.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18150601
|
R&D Systems
MAB2458 |
100 μg |
£346.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RGM-A Monoclonal specifically detects RGM-A in Human, Mouse, Chicken samples. It is validated for Western Blot.Specifications
| RGM-A | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Rat | |
| Human, Mouse, Chicken | |
| Q6PCX7 | |
| 56963 | |
| Mouse myeloma cell line NS0-derived recombinant mouse RGM-A Cys48-Gly421 Accession # Q6PCX7 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse RGM-A in direct ELISAs and Western blots. In direct ELISAs and Western blots, 100% cross-reactivity with recombinant human RGM-A and recombinant chicken RGM is observed. In Western blots, approximately 25% cross-reactivity with recombinant mouse RGM-C is observed. |
| 275407 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| RGM domain family member A, RGM domain family, member A, RGMA, RGMrepulsive guidance molecule A | |
| RGMA | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title