missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Chicken RGM-A Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB2458-SP
This item is not returnable.
View return policy
Description
RGM-A Monoclonal specifically detects RGM-A in Human, Mouse, Chicken samples. It is validated for Western Blot.
Specifications
| RGM-A | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q6PCX7 | |
| RGMA | |
| Mouse myeloma cell line NS0-derived recombinant mouse RGM-A Cys48-Gly421 Accession # Q6PCX7 | |
| 25 μg | |
| Primary | |
| Detects mouse RGM-A in direct ELISAs and Western blots. In direct ELISAs and Western blots, 100% cross-reactivity with recombinant human RGM-A and recombinant chicken RGM is observed. In Western blots, approximately 25% cross-reactivity with recombinant mouse RGM-C is observed. | |
| Human, Mouse, Chicken | |
| Purified |
| Western Blot | |
| 275407 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| RGM domain family member A, RGM domain family, member A, RGMA, RGMrepulsive guidance molecule A | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 56963 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction