missing translation for 'onlineSavingsMsg'
Learn More

Human/Mouse CD11b Rat, V450, Clone: M1/70, enQuireBio™

Rat Monoclonal Antibody

Brand:  EnquireBio QAB22-V450-25ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15941688

  • £118.00 / 25µg
Estimated Shipment: 20-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Human/Mouse CD11b
Monoclonal
0.2mg/mL
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2
Protein A/G Purified
RUO
Store at 2-4°C.
Flow Cytometry
M1/70
Violet Fluor 450 (V450)
Rat
25 μg
Primary
IgG2b κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers