missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse CD11b Rat, V450, Clone: M1/70, enQuireBio™
Rat Monoclonal Antibody
£118.00 - £256.00
Specifications
Antigen | Human/Mouse CD11b |
---|---|
Clone | M1/70 |
Concentration | 0.2mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15941688
|
EnquireBio
QAB22-V450-25UG |
25 μg |
£118.00
25µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15931688
|
EnquireBio
QAB22-V450-100UG |
100 μg |
£256.00
100µg |
Estimated Shipment: 07-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Human/Mouse CD11b | |
0.2mg/mL | |
Monoclonal | |
Rat | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
Primary | |
Store at 2-4°C. |
M1/70 | |
Flow Cytometry | |
Violet Fluor 450 (V450) | |
RUO | |
IgG2b κ | |
Protein A/G Purified |