missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse BOC Antibody, R&D Systems™

Mouse Monoclonal Antibody
£168.00 - £369.00
Specifications
| Antigen | BOC |
|---|---|
| Clone | 273729 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18709028
![]() |
R&D Systems
MAB20361-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18162601
![]() |
R&D Systems
MAB20361 |
100 μg |
£369.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BOC Monoclonal specifically detects BOC in Human, Mouse samples. It is validated for Western Blot.Specifications
| BOC | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse | |
| Q9BWV1 | |
| 91653 | |
| Mouse myeloma cell line NS0-derived recombinant human BOC Asp31-Asp852 Accession # Q9BWV1 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human and mouse BOC in direct ELISAs and Western blots. |
| 273729 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Boc homolog (mouse), brother of CDO, Protein BOC | |
| BOC | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title