missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse BOC Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB20361
This item is not returnable.
View return policy
Description
BOC Monoclonal specifically detects BOC in Human, Mouse samples. It is validated for Western Blot.
Specifications
| BOC | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q9BWV1 | |
| BOC | |
| Mouse myeloma cell line NS0-derived recombinant human BOC Asp31-Asp852 Accession # Q9BWV1 | |
| 100 μg | |
| Primary | |
| Detects human and mouse BOC in direct ELISAs and Western blots. | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| 273729 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Boc homolog (mouse), brother of CDO, Protein BOC | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 91653 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction