missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse BMP-10 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 6 publications
£181.00 - £1830.00
Specifications
| Antigen | BMP-10 |
|---|---|
| Clone | 462732 |
| Dilution | Western Blot 1 ug/mL, Neutralization 0.1-0.4 ug/mL |
| Applications | Western Blot, Neutralization |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18752589
![]() |
R&D Systems
MAB2926-SP |
25 μg |
£181.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18707809
![]() |
R&D Systems
MAB2926-100 |
100 μg |
£451.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18712878
![]() |
R&D Systems
MAB2926-500 |
500 μg |
£1830.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BMP-10 Monoclonal specifically detects BMP-10 in Human, Mouse samples. It is validated for Western Blot, Neutralization.Specifications
| BMP-10 | |
| Western Blot 1 ug/mL, Neutralization 0.1-0.4 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BMP10, BMP-10, bone morphogenetic protein 10, MGC126783 | |
| BMP10 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 462732 | |
| Western Blot, Neutralization | |
| Unconjugated | |
| Mouse | |
| Human, Mouse | |
| O95393 | |
| 27302 | |
| Chinese hamster ovary cell line CHO-derived recombinant human BMP-10 Asn317-Arg424 Accession # O95393 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human BMP-10 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) BMP-1.1, 2, 3, 3b, 4, 5, 6, 7, 9, 15, or the pro region of rhBMP-10 is observed. In direct ELISAs, approximately 90% cross-reactivity with recombinant mouse BMP-10 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title