missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse BMP-10 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 6 publications
Brand: R&D Systems MAB2926-100
This item is not returnable.
View return policy
Description
BMP-10 Monoclonal specifically detects BMP-10 in Human, Mouse samples. It is validated for Western Blot, Neutralization.
Specifications
| BMP-10 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BMP10, BMP-10, bone morphogenetic protein 10, MGC126783 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 27302 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
| Western Blot, Neutralization | |
| 462732 | |
| Western Blot 1 ug/mL, Neutralization 0.1-0.4 ug/mL | |
| O95393 | |
| BMP10 | |
| Chinese hamster ovary cell line CHO-derived recombinant human BMP-10 Asn317-Arg424 Accession # O95393 | |
| 100 μg | |
| Primary | |
| Detects human BMP-10 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant human (rh) BMP-1.1, 2, 3, 3b, 4, 5, 6, 7, 9, 15, or the pro region of rhBMP-10 is observed. In direct ELISAs, approximately 90% cross-reactivity with recombinant mouse BMP-10 is observed. | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction