missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MMP-13 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£184.00 - £1840.00
Specifications
| Antigen | MMP-13 |
|---|---|
| Clone | 87512 |
| Dilution | Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunoprecipitation 25 ug/mL, Immunoaffinity Purification, Immunocytochemistry 8-25 ug/mL |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation, Affinity Purification, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18755158
![]() |
R&D Systems
MAB511-SP |
25 μg |
£184.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18753308
![]() |
R&D Systems
MAB511-100 |
100 μg |
£466.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18725538
![]() |
R&D Systems
MAB511-500 |
500 μg |
£1840.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MMP-13 Monoclonal specifically detects MMP-13 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunoaffinity Purification, Immunocytochemistry.Specifications
| MMP-13 | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunoprecipitation 25 ug/mL, Immunoaffinity Purification, Immunocytochemistry 8-25 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| P45452 | |
| 4322 | |
| Chinese hamster ovary cell line CHO-derived recombinant human MMP-13 Leu20-Cys471 Accession # P45452 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human MMP-13 in direct ELISAs and Western blots. Detects both pro and active forms of human MMP-13 in Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human MMP-1, -2, -3, -7, -8, -9, -10, or -12 is observed. |
| 87512 | |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Affinity Purification, Immunocytochemistry | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CLG 3, CLG3EC 3.4.24, collagenase 3, EC 3.4.24.-, EC 3.4.24.22, EC 3.4.24.24, EC 3.4.24.35, EC 3.4.24.65, EC 3.4.24.7, MANDP1, matrix metallopeptidase 13 (collagenase 3), matrix metalloproteinase 13 (collagenase 3), Matrix metalloproteinase-13, MMP13, MMP-13 | |
| MMP13 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title