missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MMP-13 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB511-100
This item is not returnable.
View return policy
Description
MMP-13 Monoclonal specifically detects MMP-13 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunoaffinity Purification, Immunocytochemistry.
Specifications
| MMP-13 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CLG 3, CLG3EC 3.4.24, collagenase 3, EC 3.4.24.-, EC 3.4.24.22, EC 3.4.24.24, EC 3.4.24.35, EC 3.4.24.65, EC 3.4.24.7, MANDP1, matrix metallopeptidase 13 (collagenase 3), matrix metalloproteinase 13 (collagenase 3), Matrix metalloproteinase-13, MMP13, MMP-13 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4322 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Affinity Purification, Immunocytochemistry | |
| 87512 | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunoprecipitation 25 ug/mL, Immunoaffinity Purification, Immunocytochemistry 8-25 ug/mL | |
| P45452 | |
| MMP13 | |
| Chinese hamster ovary cell line CHO-derived recombinant human MMP-13 Leu20-Cys471 Accession # P45452 | |
| 100 μg | |
| Primary | |
| Detects human MMP-13 in direct ELISAs and Western blots. Detects both pro and active forms of human MMP-13 in Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human MMP-1, -2, -3, -7, -8, -9, -10, or -12 is observed. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction