missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MMP-12 Catalytic Domain Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 8 publications
Brand: R&D Systems MAB919
This item is not returnable.
View return policy
Description
MMP-12 Monoclonal specifically detects MMP-12 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunoaffinity Purification.
Specifications
| MMP-12 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| MMP12 | |
| Chinese hamster ovary cell line CHO-derived recombinant human MMP-12 | |
| 500 μg | |
| Primary | |
| Detects human MMP-12 Catalytic Domain. Does not recognize the C-terminal hemopexin-like domain of human MMP-12. Does not cross-react with recombinant human MMP-1, -2, -3, -7, -8, -9, -10, or -13. | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Affinity Purification | |
| 82902 | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunoprecipitation 25 ug/mL, Immunoaffinity Purification | |
| EC 3.4.24, hME, HMEEC 3.4.24.65, Macrophage elastase, macrophage metalloelastase, matrix metallopeptidase 12 (macrophage elastase), matrix metalloproteinase 12 (macrophage elastase), Matrix metalloproteinase-12, ME, MGC138506, MME, MMP12, MMP-12 | |
| Mouse | |
| Protein A or G purified from ascites | |
| RUO | |
| 4321 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido