missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MMP-12 Catalytic Domain Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 8 publications
£168.00 - £500.00
Specifications
| Antigen | MMP-12 |
|---|---|
| Clone | 82902 |
| Dilution | Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunoprecipitation 25 ug/mL, Immunoaffinity Purification |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation, Affinity Purification |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18792828
![]() |
R&D Systems
MAB919-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18061312
![]() |
R&D Systems
MAB919 |
500 μg |
£500.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MMP-12 Monoclonal specifically detects MMP-12 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunoaffinity Purification.Specifications
| MMP-12 | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunoprecipitation 25 ug/mL, Immunoaffinity Purification | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 4321 | |
| Chinese hamster ovary cell line CHO-derived recombinant human MMP-12 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human MMP-12 Catalytic Domain. Does not recognize the C-terminal hemopexin-like domain of human MMP-12. Does not cross-react with recombinant human MMP-1, -2, -3, -7, -8, -9, -10, or -13. |
| 82902 | |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Affinity Purification | |
| Unconjugated | |
| Mouse | |
| Human | |
| EC 3.4.24, hME, HMEEC 3.4.24.65, Macrophage elastase, macrophage metalloelastase, matrix metallopeptidase 12 (macrophage elastase), matrix metalloproteinase 12 (macrophage elastase), Matrix metalloproteinase-12, ME, MGC138506, MME, MMP12, MMP-12 | |
| MMP12 | |
| IgG2b | |
| Protein A or G purified from ascites | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title