missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MFRP Antibody, R&D Systems™

Rat Monoclonal Antibody
£146.00 - £346.00
Specifications
| Antigen | MFRP |
|---|---|
| Clone | 291204 |
| Concentration | LYOPH |
| Dilution | Immunohistochemistry 8-25 ug/mL |
| Applications | Immunohistochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18710257
![]() |
R&D Systems
MAB19151-SP |
25 μg |
£146.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18106131
|
R&D Systems
MAB19151 |
100 μg |
£346.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MFRP Monoclonal specifically detects MFRP in Human samples. It is validated for Immunohistochemistry.Specifications
| MFRP | |
| LYOPH | |
| Immunohistochemistry | |
| Unconjugated | |
| Rat | |
| Human | |
| Q9BY79 | |
| 83552 | |
| Mouse myeloma cell line NS0-derived recombinant human MFRP Ser101-Pro579 Accession # Q9BY79 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human MFRP in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse (rm) Frizzled-1, -2, -3, -4, -7, -8, -9, recombinant human Frizzled-5, -6, -10, or rmMFRP is observed. |
| 291204 | |
| Immunohistochemistry 8-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| MCOP5, membrane frizzled-related protein, Membrane-type frizzled-related protein, MGC32938, NNO2, RD6 | |
| MFRP | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title