missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MFRP Antibody, R&D Systems™
Rat Monoclonal Antibody
Brand: R&D Systems MAB19151
This item is not returnable.
View return policy
Description
MFRP Monoclonal specifically detects MFRP in Human samples. It is validated for Immunohistochemistry.
Specifications
| MFRP | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry 8-25 ug/mL | |
| Q9BY79 | |
| MFRP | |
| Mouse myeloma cell line NS0-derived recombinant human MFRP Ser101-Pro579 Accession # Q9BY79 | |
| 100 μg | |
| Primary | |
| Detects human MFRP in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse (rm) Frizzled-1, -2, -3, -4, -7, -8, -9, recombinant human Frizzled-5, -6, -10, or rmMFRP is observed. | |
| Human | |
| Purified |
| Immunohistochemistry | |
| 291204 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| MCOP5, membrane frizzled-related protein, Membrane-type frizzled-related protein, MGC32938, NNO2, RD6 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 83552 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction