missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MFG-E8 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£167.00 - £421.00
Specifications
| Antigen | MFG-E8 |
|---|---|
| Clone | 278901 |
| Concentration | LYOPH |
| Dilution | Western Blot 1 ug/mL, Simple Western 50 ug/mL |
| Applications | Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18745078
![]() |
R&D Systems
MAB2767-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18130791
|
R&D Systems
MAB2767 |
100 μg |
£421.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MFG-E8 Monoclonal specifically detects MFG-E8 in Human samples. It is validated for Western Blot, Simple Western.Specifications
| MFG-E8 | |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q08431 | |
| 4240 | |
| Mouse myeloma cell line NS0-derived recombinant human MFG-E8 Leu24-Cys387 Accession # Q08431 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human MFG-E8 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human (rh) C1r, rhC1s, rhC2, or rmMFG-E8. |
| 278901 | |
| Western Blot 1 ug/mL, Simple Western 50 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BA46, Breast epithelial antigen BA46, EDIL1, hP47, HsT19888, lactadherin, Lactahedrin, Medin, MFG1, MFGE8, MFG-E8, MFGM, Milk fat globule-EGF factor 8, milk fat globule-EGF factor 8 protein, O-acetyl disialoganglioside synthase, OAcGD3S, SED1, SPAG10, sperm associated antigen 10, sperm surface protein hP47 | |
| MFGE8 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title