missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MFG-E8 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB2767-SP
This item is not returnable.
View return policy
Description
MFG-E8 Monoclonal specifically detects MFG-E8 in Human samples. It is validated for Western Blot, Simple Western.
Specifications
| MFG-E8 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Simple Western 50 ug/mL | |
| Q08431 | |
| MFGE8 | |
| Mouse myeloma cell line NS0-derived recombinant human MFG-E8 Leu24-Cys387 Accession # Q08431 | |
| 25 μg | |
| Primary | |
| Detects human MFG-E8 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant human (rh) C1r, rhC1s, rhC2, or rmMFG-E8. | |
| Human | |
| Purified |
| Western Blot | |
| 278901 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BA46, Breast epithelial antigen BA46, EDIL1, hP47, HsT19888, lactadherin, Lactahedrin, Medin, MFG1, MFGE8, MFG-E8, MFGM, Milk fat globule-EGF factor 8, milk fat globule-EGF factor 8 protein, O-acetyl disialoganglioside synthase, OAcGD3S, SED1, SPAG10, sperm associated antigen 10, sperm surface protein hP47 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4240 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction