missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MFAP3 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£161.00 - £537.00
Specifications
| Antigen | MFAP3 |
|---|---|
| Clone | 1059216 |
| Dilution | Immunohistochemistry 5-25 ug/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18702737
![]() |
R&D Systems
MAB10798-SP |
25 μg |
£161.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18720247
|
R&D Systems
MAB10798-100 |
100 μg |
£537.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MFAP3 Monoclonal antibody specifically detects MFAP3 in Human samples. It is validated for ImmunohistochemistrySpecifications
| MFAP3 | |
| Immunohistochemistry 5-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| DKFZp586F0219, microfibril-associated glycoprotein 3, microfibrillar-associated protein 3 | |
| Human embryonic kidney cell, HEK293-derived human MFAP3, Leu22-Met147, Accession # P55082 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. |
| 1059216 | |
| Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| P55082 | |
| 4238 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title