Learn More
Human MFAP3 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB10798-100
 View more versions of this product
Product Code. 18720247
  • £537.00 / 100µg
Description
MFAP3 Monoclonal antibody specifically detects MFAP3 in Human samples. It is validated for Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MFAP3
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS.
DKFZp586F0219, microfibril-associated glycoprotein 3, microfibrillar-associated protein 3
Human embryonic kidney cell, HEK293-derived human MFAP3, Leu22-Met147, Accession # P55082
100 μg
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution.
IgG1
Immunohistochemistry
1059216
Immunohistochemistry 5-25 ug/mL
P55082
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
4238
Human
Purified