missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MD-2 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF1787
This item is not returnable.
View return policy
Description
MD-2 Polyclonal specifically detects MD-2 in Human samples. It is validated for Western Blot.
Specifications
| MD-2 | |
| Polyclonal | |
| Unconjugated | |
| NP_001123946 | |
| LY96 | |
| E. coli-derived recombinant human MD-2 Glu17-Asn160 Accession # NP_001123946 | |
| 100 μg | |
| Primary | |
| Detects human MD-2 in direct ELISAs and Western blots. In these formats, less than 1% cross-reactivity with recombinant human MD-1 is observed. | |
| Human | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| ESOP1, ESOP-1, Ly-96, LY96, lymphocyte antigen 96, MD2, MD-2, myeloid differentiation protein-2, Protein MD-2 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 23643 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction