missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MD-2 Antibody, R&D Systems™

Sheep Polyclonal Antibody
£168.00 - £408.00
Specifications
| Antigen | MD-2 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18729057
![]() |
R&D Systems
AF1787-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18042692
![]() |
R&D Systems
AF1787 |
100 μg |
£408.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MD-2 Polyclonal specifically detects MD-2 in Human samples. It is validated for Western Blot.Specifications
| MD-2 | |
| Western Blot 0.1 ug/mL | |
| Polyclonal | |
| Sheep | |
| Human | |
| ESOP1, ESOP-1, Ly-96, LY96, lymphocyte antigen 96, MD2, MD-2, myeloid differentiation protein-2, Protein MD-2 | |
| LY96 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| NP_001123946 | |
| 23643 | |
| E. coli-derived recombinant human MD-2 Glu17-Asn160 Accession # NP_001123946 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human MD-2 in direct ELISAs and Western blots. In these formats, less than 1% cross-reactivity with recombinant human MD-1 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title