missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MD-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£167.00 - £492.00
Specifications
| Antigen | MD-1/LY86 |
|---|---|
| Clone | 153014 |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18723788
![]() |
R&D Systems
MAB925-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18110871
![]() |
R&D Systems
MAB925 |
500 μg |
£492.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MD-1/LY86 Monoclonal specifically detects MD-1/LY86 in Human samples. It is validated for Western Blot.Specifications
| MD-1/LY86 | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Ly-86, LY86, lymphocyte antigen 86, MD1, MMD-1, Protein MD-1, RP105-associated | |
| LY86 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 153014 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| O95711 | |
| 9450 | |
| Mouse myeloma cell line NS0-derived recombinant human MD-1 Gly21-Ser162 Accession # O95711 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human MD-1 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant mouse MD-1 is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title