missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MD-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB925
This item is not returnable.
View return policy
Description
MD-1/LY86 Monoclonal specifically detects MD-1/LY86 in Human samples. It is validated for Western Blot.
Specifications
| MD-1/LY86 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Ly-86, LY86, lymphocyte antigen 86, MD1, MMD-1, Protein MD-1, RP105-associated | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 9450 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot | |
| 153014 | |
| Western Blot 1 ug/mL | |
| O95711 | |
| LY86 | |
| Mouse myeloma cell line NS0-derived recombinant human MD-1 Gly21-Ser162 Accession # O95711 | |
| 500 μg | |
| Primary | |
| Detects human MD-1 in direct ELISAs and Western blots. In Western blots, no cross-reactivity with recombinant mouse MD-1 is observed. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction