missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MCEMP1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
£168.00 - £491.00
Specifications
| Antigen | MCEMP1 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, CyTOF |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18745817
![]() |
R&D Systems
AF4994-SP |
25 μg |
£168.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18079561
|
R&D Systems
AF4994 |
100 μg |
£491.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MCEMP1 Polyclonal specifically detects MCEMP1 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
| MCEMP1 | |
| Western Blot 0.1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| Polyclonal | |
| Sheep | |
| Human | |
| C19orf59, chromosome 19 open reading frame 59, mast cell-expressed membrane protein 1, MCEMP1, MGC132456 | |
| C19ORF59 | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, Flow Cytometry, CyTOF | |
| Unconjugated | |
| RUO | |
| Q8IX19 | |
| 199675 | |
| Mouse myeloma cell line NS0-derived recombinant human MCEMP1 Val106-Gln187 Accession # Q8IX19 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human MCEMP1 in direct ELISAs and Western blots. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title