missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MCEMP1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF4994-SP
This item is not returnable.
View return policy
Description
MCEMP1 Polyclonal specifically detects MCEMP1 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| MCEMP1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| C19orf59, chromosome 19 open reading frame 59, mast cell-expressed membrane protein 1, MCEMP1, MGC132456 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 199675 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Flow Cytometry, CyTOF | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| Q8IX19 | |
| C19ORF59 | |
| Mouse myeloma cell line NS0-derived recombinant human MCEMP1 Val106-Gln187 Accession # Q8IX19 | |
| 25 μg | |
| Primary | |
| Detects human MCEMP1 in direct ELISAs and Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction