missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MASTL Antibody, R&D Systems™

Goat Antibody
£155.00 - £491.00
Specifications
| Antigen | MASTL |
|---|---|
| Dilution | Immunocytochemistry 5-15 μg;/mL |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18765796
![]() |
R&D Systems
AF10022-SP |
25 μg |
£155.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18728806
![]() |
R&D Systems
AF10022-100 |
100 μg |
£491.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MASTL specifically detects MASTL in Human samples. It is validated for Immunocytochemistry.Specifications
| MASTL | |
| Immunocytochemistry | |
| Unconjugated | |
| Goat | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| FLJ14813EC 2.7.11.1, GREATWALL, GW, GWL, hGWL, MAST-L, microtubule associated serine/threonine kinase-like, Microtubule-associated serine/threonine-protein kinase-like, RP11-85G18.2, THC2, THC2greatwall protein kinase | |
| E. coli-derived recombinant human MASTL., Lys39-Lys158, Accession No. Q96GX5 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects human MASTL in direct ELISA. |
| Immunocytochemistry 5-15 μg;/mL | |
| Polyclonal | |
| Lyophilized | |
| Human | |
| Q96GX5 | |
| 84930.0 | |
| IgG | |
| Antigen Affinity-purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title