missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MASTL Antibody, R&D Systems™

Goat Antibody
Brand: R&D Systems AF10022-SP
This item is not returnable.
View return policy
Description
MASTL specifically detects MASTL in Human samples. It is validated for Immunocytochemistry.
Specifications
| MASTL | |
| Polyclonal | |
| Immunocytochemistry 5-15 μg;/mL | |
| Q96GX5 | |
| Goat | |
| Antigen Affinity-purified | |
| Primary | |
| Detects human MASTL in direct ELISA. | |
| Human | |
| Lyophilized |
| Immunocytochemistry | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| FLJ14813EC 2.7.11.1, GREATWALL, GW, GWL, hGWL, MAST-L, microtubule associated serine/threonine kinase-like, Microtubule-associated serine/threonine-protein kinase-like, RP11-85G18.2, THC2, THC2greatwall protein kinase | |
| E. coli-derived recombinant human MASTL., Lys39-Lys158, Accession No. Q96GX5 | |
| 25 μg | |
| 84930.0 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction