missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MAP2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£177.00 - £387.00
Specifications
| Antigen | MAP2 |
|---|---|
| Clone | 885232 |
| Dilution | Immunohistochemistry 0.5-25 ug/mL, Immunocytochemistry 0.5-25 ug/mL |
| Applications | Immunohistochemistry, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18722858
![]() |
R&D Systems
MAB8304-SP |
25 μg |
£177.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18132381
|
R&D Systems
MAB8304 |
100 μg |
£387.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MAP2 Monoclonal specifically detects MAP2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry.Specifications
| MAP2 | |
| Immunohistochemistry 0.5-25 ug/mL, Immunocytochemistry 0.5-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in TBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DKFZp686I2148, MAP-2, MAP2A, MAP2B, MAP2C, Microtubule Associated Protein 2, microtubule-associated protein 2, MTAP2 | |
| MAP2 | |
| IgG3 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 885232 | |
| Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| P11137 | |
| 4133 | |
| E. coli-derived recombinant human MAP2 Gly1689-Lys1824 Accession # P11137 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human MAP2 in direct ELISAs. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title