missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MAP2 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB8304
This item is not returnable.
View return policy
Description
MAP2 Monoclonal specifically detects MAP2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry.
Specifications
| MAP2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in TBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DKFZp686I2148, MAP-2, MAP2A, MAP2B, MAP2C, Microtubule Associated Protein 2, microtubule-associated protein 2, MTAP2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4133 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG3 |
| Immunohistochemistry, Immunocytochemistry | |
| 885232 | |
| Immunohistochemistry 0.5-25 ug/mL, Immunocytochemistry 0.5-25 ug/mL | |
| P11137 | |
| MAP2 | |
| E. coli-derived recombinant human MAP2 Gly1689-Lys1824 Accession # P11137 | |
| 100 μg | |
| Primary | |
| Detects human MAP2 in direct ELISAs. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction